Order Entry
Austria
Orders LinkContactUsLinkComponent
Liquid chromatography sorbents, Polyamide 6 | Avantor

Liquid chromatography sorbents, Polyamide 6

Polyamide CC 6, with particle size of 0,10 - 0,30 mm, 5 kg
Catalog # MANA815600.5
Supplier:   MACHEREY-NAGEL
Liquid chromatography sorbents, Polyamide 6
Catalog # MANA815600.5
Supplier:  MACHEREY-NAGEL
CAS Number:  
Liquid chromatography sorbents, Polyamide 6
Liquid chromatography sorbents, Polyamide 6

Polyamide CC 6, with particle size of 0,10 - 0,30 mm, 5 kg

CAS Number:  
Price
1 885,00 €
Log in for availability
1 * 5 kg
Product Details & Documents

Polyamide 6 adsorbents for column chromatography.

  • Polyamide 6 (ε-polycaprolactam)
  • Chromatographic quality
  • Separation mechanism mainly based on hydrogen bonds

Recommended application: separation of phenolic compounds (e.g., isolation of natural products), carboxylic acids and aromatic nitro compounds.

Dokumente

Specifications

Specifications

  • Pk:
    5 kg
  • Description:
    Polyamide CC 6, with particle size of 0,10 - 0,30 mm

Specifications