Order Entry
Austria
Orders LinkContactUsLinkComponent
 

VISOCOLOR® Colorimetric Test, Nitrite

VISOCOLOR® alpha Nitrite, Reagents for 200 tests and color chart in a blister, 0.05 • 0.10 • 0.25 • 0.5 • 1.0 mg/L (NO₂⁻), Sulfanilic acid/1, Visual, 92×153×35 mm (including packaging), Hazardous material: Yes, 200, 1 KIT
Catalog # MANA935066
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Nitrite
Catalog # MANA935066
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, Nitrite
VISOCOLOR® Colorimetric Test, Nitrite

VISOCOLOR® alpha Nitrite, Reagents for 200 tests and color chart in a blister, 0.05 • 0.10 • 0.25 • 0.5 • 1.0 mg/L (NO₂⁻), Sulfanilic acid/1, Visual, 92×153×35 mm (including packaging), Hazardous material: Yes, 200, 1 KIT

CAS Number:  
Price
30,80 €
Log in for availability
1 * 1 KIT
Specifications

Specifications

  • Chemical method:
    Sulfanilic acid/1
  • Pk:
    1 KIT
  • Description:
    VISOCOLOR® alpha Nitrite
  • Dimensions:
    92×153×35 mm (including packaging)
  • Type:
    Hazardous material: Yes
  • For unit:
    Visual
  • Tests per kit:
    200
  • Range:
    0.05 • 0.10 • 0.25 • 0.5 • 1.0 mg/L (NO₂⁻)
  • Contents:
    Reagents for 200 tests and color chart in a blister
  • Cat. No.:
    MANA935066
  • Platform:
    Colorimetric test kits
  • Model:
    MANA935066
  • Shelf life (months):
    18
  • Storage temperature:
    15…25 °C
  • Weight:
    41,25 g (including packaging)

Specifications