VISOCOLOR® Colorimetric Test, Nitrite
VISOCOLOR® ECO Nitrite, test no.5-44; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 120 tests, color chart and accessories in a box, 0 • 0,02 • 0,03 • 0,05 • 0,07 • 0,1 • 0,2 • 0,3 • 0,5 mg/L(NO₂⁻); 0,02 - 0,50 mg/L (NO₂⁻), Sulfanilic acid/1, PF-12Plus; PF-3 Fish; Advance; Visual, 168×112×40 mm (including packaging), Hazardous material: Yes, 120, 1 KIT
Anbieter: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Reagents for the determination of nitrite in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Documents
Specifications
Specifications
- Chemical method:Sulfanilic acid/1
- Pk:1 KIT
- Description:VISOCOLOR® ECO Nitrite, test no.5-44; Measuring range on PF-12Plus. Range on other photometers can be different
- Dimensions:168×112×40 mm (including packaging)
- Type:Hazardous material: Yes
- For unit:PF-12Plus; PF-3 Fish; Advance; Visual
- Tests per kit:120
- Range:0 • 0,02 • 0,03 • 0,05 • 0,07 • 0,1 • 0,2 • 0,3 • 0,5 mg/L(NO₂⁻); 0,02 - 0,50 mg/L (NO₂⁻)
- Contents:Reagents for 120 tests, color chart and accessories in a box
- Cat. No.:MANA931044
- Platform:Colorimetric test kits
- Model:MANA931044
- Shelf life (months):18
- Storage temperature:15…25 °C
- Weight:189,74 g (including packaging)



