Empty solid phase extraction columns, CHROMABOND®
Empty PP columns with PE frits, one filter element is already inserted in the PP column, 30 ml
Catalog # MANA730380
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Empty and reservoir columns with accessories for solid phase extraction.
- Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
- Reservoir columns from PP for application of medium-size samples
- Accessories for SPE columns (filter elements, adapters)
Specifications
- Capacity:30 ml
- Description:Empty PP columns with PE frits, one filter element is already inserted in the PP column
- Seq:05
Accessories
Product Information
- Catalog #AvailabilityPrice
- PE frits for LV PP columns 15 ml, for 100 mg adsorbent weightPE frits for LV PP columns 15 ml, for 100 mg adsorbent weightMANA732019Log in for availability1 * 250 ST - 132,00 €
Product Information
- Catalog #AvailabilityPrice
- PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weightPE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weightMANA732020Log in for availability1 * 250 ST - 185,00 €
Product Information
- Catalog #AvailabilityPrice
- Adapters (PP) for PP columns (1, 3 and 6 ml)
Product Information
- Catalog #AvailabilityPrice
- Adapters (PVDF) for glass columns (3 and 6 ml)Adapters (PVDF) for glass columns (3 and 6 ml)MANA730105Log in for availability1 * 10 ST - 200,00 €
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 70 ml
Product Information
- Catalog #AvailabilityPrice
- Glass fiber frits for glass columns, 6 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 15 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 45 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 150 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 30 ml
Product Information
- Catalog #AvailabilityPrice
- Adapters (PE) for PP columns (30 and 70 ml)
Product Information
- Catalog #AvailabilityPrice
- Adapters (PE) for PP columns (15, 45 and 70 ml)Adapters (PE) for PP columns (15, 45 and 70 ml)MANA730385Log in for availability1 * 10 ST - 125,00 €
Product Information
- Catalog #AvailabilityPrice
- Glass fiber frits for glass columns, 3 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 1 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 6 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 3 ml



