Order Entry
Austria
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

60×180 mm, 2,0 mm
Catalog # MANA645039
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Catalog # MANA645039
Supplier:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles

60×180 mm, 2,0 mm

Price
261,00 €
Einloggen für Verfügbarkeit
1 * 25 ST
Product Details & Documents

Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.

  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Spezifikationen
  • Int.Ø×H:
    60×180 mm
  • Seq:
    40
  • Thickness:
    2,0 mm