Order Entry
Austria
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

57×315 mm, 2,0 mm
Catalog # MANA645038
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Catalog # MANA645038
Supplier:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles

57×315 mm, 2,0 mm

Price
385,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.

  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifications
  • Int.Ø×H:
    57×315 mm
  • Seq:
    39
  • Thickness:
    2,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.