Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Extraction thimbles

30× 60 mm, 1,5 mm
Catalog # MANA645019
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Catalog # MANA645019
Supplier:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles

30× 60 mm, 1,5 mm

Price
97,70 €
Log in for availability
1 * 25 ST
Product Details & Documents

Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.

  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifications
  • Int.Ø×H:
    30× 60 mm
  • Seq:
    17
  • Thickness:
    1,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.