Order Entry
Austria
Orders LinkContactUsLinkComponent
Cycloheximide

Cycloheximide

Glass bottle for solids, 25 g
Catalog # A0879.0025
Toxic
Health
Environment
Supplier:   PanReac AppliChem
Cycloheximide
Catalog # A0879.0025
Supplier:  PanReac AppliChem
CAS Number:  66-81-9
Cycloheximide
Cycloheximide

Glass bottle for solids, 25 g

CAS Number:  66-81-9
Price
1 050,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications