Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 500 mL
Catalog # A4892.0500
Supplier:   PanReac AppliChem
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Catalog # A4892.0500
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 500 mL

CAS Number:  
Price
50,10 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    EDTA buffer solution pH 8,0 (0,5 mol/l)

Specifications