Order Entry
Austria
Orders LinkContactUsLinkComponent
Agar

Agar

Glass bottle for solids, 500 g
Catalog # APPCA0949.0500
Supplier:   PanReac AppliChem
Agar
Catalog # APPCA0949.0500
Supplier:  PanReac AppliChem
CAS Number:  9002-18-0
Agar
Agar

Glass bottle for solids, 500 g

CAS Number:  9002-18-0
Price
159,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications