Orders LinkContactUsLinkComponent
Base materials
Base materials
Tryptone pancreatic digest of casein, Glass bottle for solids, 1 kg
Catalog # MOBO12111-1
Marketsource
Supplier:   MO BIO
Base materials
Catalog # MOBO12111-1
Supplier:  MO BIO
Marketsource
CAS Number:  
Base materials
Tryptone pancreatic digest of casein, Glass bottle for solids, 1 kg
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Bacteriological grade enzymatic digest of casein.
  • BSE free
Specifications
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Description:
    Tryptone pancreatic digest of casein