Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonium Chloride Solution

Ammonium Chloride Solution

Reagent for lysis of red blood cells, 500 mL
Catalog # STMC07850
Marketsource
Ammonium Chloride Solution
Catalog # STMC07850
Marketsource
CAS Number:  
Ammonium Chloride Solution
Ammonium Chloride Solution

Reagent for lysis of red blood cells, 500 mL

CAS Number:  
Price
271,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Ammonium chloride solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen or bone marrow cells. It is buffered and optimised for gentle lysis of erythrocytes, with minimal effect on leukocytes. It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis.

Species: Human, mouse, non human primate, other, rat

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Reagent for lysis of red blood cells

Specifications