Orders LinkContactUsLinkComponent
Forceps
Forceps
Sharp, curved, 125 mm
Catalog # 232-0037
Supplier:   MURRAY S
Forceps
Catalog # 232-0037
Supplier:  MURRAY S
Forceps
Forceps
Sharp, curved, 125 mm
Price
7,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel. Dissecting forceps with guide pin, which helps to keep the fine tips aligned.
  • Sharp, narrow flutes
  • Curved
Specifications
  • Type:
    Sharp, curved
  • Length:
    125 mm