Order Entry
Austria
Orders LinkContactUsLinkComponent
Handheld refractometers

Handheld refractometers

Handheld refractometer, R-5000, 1,333 - 1,520 nD, 0,001 nD
Catalog # 635-0134
Supplier:   Atago
Handheld refractometers
Catalog # 635-0134
Supplier:  Atago
Handheld refractometers
Handheld refractometers

Handheld refractometer, R-5000, 1,333 - 1,520 nD, 0,001 nD

Price
495,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These handheld refractometers provide three stages of scale - Calibrated with water.

Specifications
  • Model:
    R-5000
  • Range:
    1,333 - 1,520 nD
  • Resolution:
    0,001 nD
  • Description:
    Handheld refractometer
  • Cat. No.:
    635-0134
  • W×D×H:
    40×40×20 mm
  • Weight:
    600 g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.