Order Entry
Austria
Orders LinkContactUsLinkComponent
 

VWR® Buffer Standard Solutions

7,00, ±0,03, 25 °C, Plastic bottle, 500 mL
Catalog # E459-500ML
Supplier:   VWR Chemicals
VWR® Buffer Standard Solutions
Catalog # E459-500ML
Supplier:  VWR Chemicals
CAS Number:  
VWR® Buffer Standard Solutions
VWR® Buffer Standard Solutions

7,00, ±0,03, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
78,40 €
Log in for availability
1 * 500 mL
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,03
  • pH value:
    7,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.