Express Eingabe
Austria
AuftragsverfolgungKontakt
Specific gravity cups, calibrated
Specific gravity cups, calibrated
Stainless steel, 50 ml
Catalog # SIMEVF2100
Supplier:   Industrial Physics
Specific gravity cups, calibrated
Catalog # SIMEVF2100
Supplier:  Industrial Physics
Specific gravity cups, calibrated
Specific gravity cups, calibrated
Stainless steel, 50 ml
Price
395,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
  • Density is defined as weight per unit volume at a specified temperature
  • Available in anodised aluminium or stainless steel
Zertifizierung: According to ISO 2811, DIN 53 217 and ASTM D 1475.
Lieferumfang: Supplied with a calibration certificate.
Documents
Specifications
  • Volumen (nom.):
    50 ml
  • Bezeichnung:
    Stainless steel
  • Seq:
    03