Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Blue
Artikelnr. WHEAW242424-A
Anbieter:   DWK Life Sciences
Dropper tips, WHEATON®
Artikelnr. WHEAW242424-A
Anbieter:  DWK Life Sciences
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Blue
Preis
491,00 €
Einloggen für Verfügbarkeit
1 * 1.000 ST
Product Details & Documents
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
 : Tips available in different colours and packaged in case of 100 or 1000.
Spezifikationen
  • Colour:
    Blue
  • Seq:
    48