Scintillation vials, PET, WHEATON®
20,00 ml, 27×61 mm, Yes, PP, Metal foil
Catalog # WHEA986741
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Manufactured from clear polyethylene terephthalate (PET) these vials have advantages over glass counting vials of low background count, economy and virtual unbreakability.
- Clarity of glass
- Can be safely incinerated
- Lightweight and virtually unbreakable
* With cap on
Specifications
- Cap included:Yes
- Cap material:PP
- Ø×H:27×61 mm
- Septum:Metal foil
- Capacity:20,00 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



