Orders LinkContactUsLinkComponent
Scintillation vials, PET, WHEATON®
Scintillation vials, PET, WHEATON®
20,00 ml, 27×61* mm, Yes, PE
Catalog # 548-2048
Supplier:   DWK Life Sciences
Scintillation vials, PET, WHEATON®
Catalog # 548-2048
Supplier:  DWK Life Sciences
Scintillation vials, PET, WHEATON®
Scintillation vials, PET, WHEATON®
20,00 ml, 27×61* mm, Yes, PE
Price
394,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
Manufactured from clear polyethylene terephthalate (PET) these vials have advantages over glass counting vials of low background count, economy and virtual unbreakability.
  • Clarity of glass
  • Can be safely incinerated
  • Lightweight and virtually unbreakable
* With cap on
Specifications
  • Cap included:
    Yes
  • Cap material:
    PE
  • Ø×H:
    27×61* mm
  • Capacity:
    20,00 ml