Calibration weights, cylindrical, OIML, class E2
1 kg, 1,6 ± mg, Cylindrical test weight
Artikelnr. 611-0896
Anbieter: Sartorius Balances
Jump to:
Product Details & Documents
Class E2 cyclindrical calibration weights.
Specifications
- Weight:1 kg
- Tolerance:1,6 ± mg
- Type:Cylindrical test weight