Order Entry
Austria
Orders LinkContactUsLinkComponent
Certified colour reference standards

Certified colour reference standards

Saybolt Colour 0, (certified to ISO 17025), 500 mL
Catalog # 633-0006
Supplier:   Lovibond Tintometer
Certified colour reference standards
Catalog # 633-0006
CAS Number:  
Certified colour reference standards
Certified colour reference standards

Saybolt Colour 0, (certified to ISO 17025), 500 mL

CAS Number:  
Price
291,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

For routine calibration of colorimeters and verification of test data.

Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Saybolt Colour 0, (certified to ISO 17025)

Specifications