Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, PBS solution 10X concentrate (phosphate buffered saline)

Buffer, PBS solution 10X concentrate (phosphate buffered saline)

PBS buffer solution 10X concentrate (phosphate buffered saline), Glass bottle, 3.000 mL
Catalog # TREV4870-500-6
Marketsource
Supplier:   TREVIGEN
Buffer, PBS solution 10X concentrate (phosphate buffered saline)
Catalog # TREV4870-500-6
Supplier:  TREVIGEN
Marketsource
CAS Number:  
Buffer, PBS solution 10X concentrate (phosphate buffered saline)

PBS buffer solution 10X concentrate (phosphate buffered saline), Glass bottle, 3.000 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 3.000 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    3.000 mL
  • Description:
    PBS buffer solution 10X concentrate (phosphate buffered saline)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.