Orders LinkContactUsLinkComponent
Hose clips, worm-threaded
Hose clips, worm-threaded
20 - 32 mm
Catalog # BURK8677-2032
Supplier:   BURKLE
Hose clips, worm-threaded
Catalog # BURK8677-2032
Supplier:  BURKLE
Hose clips, worm-threaded
Hose clips, worm-threaded
20 - 32 mm
Price
16,80 €
Log in for availability
1 * 10 ST
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Specifications
  • Spanning width:
    20 - 32 mm
  • Seq:
    05