Hose clips, worm-threaded
12 - 20 mm
Catalog # BURK8677-1220
Supplier: BURKLE
Springe zu:
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Documents
Spezifikationen
- Spanning width:12 - 20 mm
- Seq:03