Orders LinkContactUsLinkComponent
Hose clips, worm-threaded
Hose clips, worm-threaded
10 - 16 mm
Catalog # BURK8677-1016
Supplier:   BURKLE
Hose clips, worm-threaded
Catalog # BURK8677-1016
Supplier:  BURKLE
Hose clips, worm-threaded
Hose clips, worm-threaded
10 - 16 mm
Price
14,10 €
Log in for availability
1 * 10 ST
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Specifications
  • Spanning width:
    10 - 16 mm
  • Seq:
    02