Orders LinkContactUsLinkComponent
Iris scissors, Aesculap®
Perwitzschky Salival, curved, sharp/with knob, 100 mm
Catalog # AESCOM489R
Supplier:   Aesculap
Iris scissors, Aesculap®
Catalog # AESCOM489R
Supplier:  Aesculap
Iris scissors, Aesculap®
Perwitzschky Salival, curved, sharp/with knob, 100 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Perwitzschky Salival, curved, sharp/with knob
  • Length:
    100 mm
  • Seq:
    38