Orders LinkContactUsLinkComponent
Separating funnels
Separating funnels
2000 ml, 29/32 NS
Catalog # QVFLALG0050706
Supplier:   QVF LABORTECHNIK
Separating funnels
Catalog # QVFLALG0050706
Supplier:  QVF LABORTECHNIK
Separating funnels
2000 ml, 29/32 NS
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Made of borosilicate glass.
  • Stopcock with glass key
  • Without graduations
  • Excellent chemical and temperature resistance
The separating funnels are used for discontinuous shaking out of solvents.
Certifications: Complies to DIN 12 451.
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    2000 ml
  • Seq:
    05