Order Entry
Austria
Orders LinkContactUsLinkComponent
Sockets for rods without thread

Sockets for rods without thread

13,2 mm, Socket without thread, for 12 - 13 mm Ø rods, Stainless steel 18/10
  BOCH5014
 :   BOCHEM
Sockets for rods without thread
  BOCH5014
 :  BOCHEM
Sockets for rods without thread
Sockets for rods without thread

13,2 mm, Socket without thread, for 12 - 13 mm Ø rods, Stainless steel 18/10

 
20,90 €
   
1 * 1 ST
 

These sockets are available in two materials: 18/10 stainless steel or green coated steel.

Suitable for rods with a diameter of 12 - 13 mm.

 
  • Ø:
    13,2 mm
  • Description:
    Socket without thread, for 12 - 13 mm Ø rods
  • Material:
    Stainless steel 18/10
  • Seq:
    01