Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Precision forceps, smooth tips, PTFE coating, Straight, extra sharp, 120 mm
Catalog # BOCH1860
Supplier:   BOCHEM
Forceps, precision
Catalog # BOCH1860
Supplier:  BOCHEM
Forceps, precision
Forceps, precision
Precision forceps, smooth tips, PTFE coating, Straight, extra sharp, 120 mm
Price
30,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These non serrated precision forceps are available in pure stainless steel or with PTFE coating.
Specifications
  • Type:
    Straight, extra sharp
  • Description:
    Precision forceps, smooth tips, PTFE coating
  • Length:
    120 mm