Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Spoons, POLY

Oblique, 215 mm, 70×30 mm
  231-1296
 :   BOCHEM
Spoons, POLY
  231-1296
 :  BOCHEM
Spoons, POLY
Spoons, POLY

Oblique, 215 mm, 70×30 mm

 
7,60 €
   
1 * 1 ST
 

Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. Special shape for light and easy handling.

  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
 
  • Description:
    Oblique
  • Length:
    215 mm
  • Seq:
    05
  • Spoon size:
    70×30 mm