Wire baskets
270×180 mm
BOCH10056
: BOCHEM
Stainless steel.
- Spot-welded grid 8×8×1 mm
- Reinforced edges
- Stowable handle
These round baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
- Ø×H:270×180 mm
- Seq:07



