Order Entry
Austria
Orders LinkContactUsLinkComponent
Retort rods, without thread

Retort rods, without thread

12 mm, 1250 mm
Catalog # BOCH5164
Supplier:   BOCHEM
Retort rods, without thread
Catalog # BOCH5164
Supplier:  BOCHEM
Retort rods, without thread
Retort rods, without thread

12 mm, 1250 mm

Price
20,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Galvanised steel.

Specifications
  • Ø:
    12 mm
  • Length:
    1250 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.