Orders LinkContactUsLinkComponent
Stand rods, with M10 thread
12 mm, 1000 mm
  241-3160
 :   BOCHEM
Stand rods, with M10 thread
  241-3160
 :  BOCHEM
Stand rods, with M10 thread
Stand rods, with M10 thread
12 mm, 1000 mm
17,00 €
1 * 1 ST
Aluminium, bare.
  • Lightweight, tough
  • With M10 thread
  • Ø:
    12 mm
  • Length:
    1000 mm