Orders LinkContactUsLinkComponent
Tissue forceps
Tissue forceps
Tissue forceps 1:2, 130 mm
Catalog # 232-0002
Supplier:   BOCHEM
Tissue forceps
Catalog # 232-0002
Supplier:  BOCHEM
Tissue forceps
Tissue forceps
Tissue forceps 1:2, 130 mm
Price
4,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • With small teeth (1:2) at the end of the blades
  • Designed for handling tissues with minimal trauma during surgery
Specifications
  • Type:
    Tissue forceps 1:2
  • Length:
    130 mm