Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Stools

Roll stool to step on, steel, Ø 290 mm, 425 mm
  BOCH8001
 :   BOCHEM
Stools
  BOCH8001
 :  BOCHEM
Stools
Stools

Roll stool to step on, steel, Ø 290 mm, 425 mm

 
153,00 €
   
1 * 1 ST
 

Kickstool with GS safety control.

 
  • Description:
    Roll stool to step on, steel, Ø 290 mm
  • Height:
    425 mm
  • Seq:
    2
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.