Orders LinkContactUsLinkComponent
Non-return valves
Non-return valves
7,6 - 10 mm ID
Catalog # 229-3330
Supplier:   KARTELL
Non-return valves
Catalog # 229-3330
Supplier:  KARTELL
Non-return valves
Non-return valves
7,6 - 10 mm ID
Price
37,50 €
Log in for availability
1 * 10 ST
Product Details & Documents
PP, translucent. Prevent pressure loss and backflow. Maximum working pressure 2 bar and minimum working pressure  0,07 bar at room temperature (20 °C).
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Tapered ends to ensure that tubes are held firmly
Specifications
  • Tubing size:
    7,6 - 10 mm ID