Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Ammonia solution 25%, TECHNICAL

Plastic bottle, fluorinated, 5 L
Catalog # 21191.364
Corrosive
Irritant
Environment
Supplier:   VWR Chemicals
Ammonia solution 25%, TECHNICAL
Catalog # 21191.364
Supplier:  VWR Chemicals
CAS Number:  1336-21-6
Ammonia solution 25%, TECHNICAL
Ammonia solution 25%, TECHNICAL

Plastic bottle, fluorinated, 5 L

CAS Number:  1336-21-6
Price
52,60 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.907 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specification Test Results

  • Assay:
    Min. 24 - 26 %

Specifications

  • Pack type:
    Plastic bottle, fluorinated
  • Pk:
    5 L

Specifications