Dropper tips, WHEATON®
Blue
Catalog # 215-4797
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
LDPE, coloured.
- Extended controlled dropper tips
- Dispense one drop at a time
- Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
Specifications
- Colour:Blue
- Seq:49
- Cat. No.:215-4797