Orders LinkContactUsLinkComponent
Robotic tips for Tecan® | Avantor
Robotic tips for Tecan®
903-251G (wide bore), 200 µl, +, -, Black, 10 racks, 96 each
Catalog # 736-0054
   Low Manufacturing Impact
Robotic tips for Tecan®
Catalog # 736-0054
   Low Manufacturing Impact
Robotic tips for Tecan®
Robotic tips for Tecan®
903-251G (wide bore), 200 µl, +, -, Black, 10 racks, 96 each
Price
224,00 €
Log in for availability
1 * 960 ST
Product Details & Documents
High quality tips, especially intended for research in molecular biology laboratories.
  • Transparent or black PP
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Certified free of RNase, DNase, DNA and pyrogens
  • Sterile tips: Sterilised by gamma radiation
  • Wide aperture tips: Designed for applications with more viscous samples
Bestellinformation: Available in boxes of 10 racks or 50 racks (Econopak).
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    200 µl
  • Tip style:
    903-251G (wide bore)
  • Tip colour:
    Black
  • Filter:
    -
  • Cat. No.:
    736-0054
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.