Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample cuvettes

Sample cuvettes

Sample cuvettes, with rubber stopper, barcoded, 7 ml, 13 mm, Glass
Catalog # HACHLCW906
Supplier:   Hach
Sample cuvettes
Catalog # HACHLCW906
Supplier:  Hach
Sample cuvettes
Sample cuvettes

Sample cuvettes, with rubber stopper, barcoded, 7 ml, 13 mm, Glass

Price
72,70 €
Log in for availability
1 * 25 ST
Product Details & Documents

Sample cuvettes for photometric measurements.

  • Available with or without lid
  • Available reusable (made from glass) or for single use (made from plastics)
Specifications
  • Path length:
    13 mm
  • Volume:
    7 ml
  • Window material:
    Glass
  • Description:
    Sample cuvettes, with rubber stopper, barcoded
  • Seq:
    26