Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Multiscreen® Filter Plates, Opaque

GFB plate, filtration area 0,26 cm², Titanium, Opaque, 50 - 250 μl, PVDF, 1,0 µm, 96, -
Catalog # 736-0179
Supplier:   Merck
Multiscreen® Filter Plates, Opaque
Catalog # 736-0179
Supplier:  Merck
Multiscreen® Filter Plates, Opaque
Multiscreen® Filter Plates, Opaque

GFB plate, filtration area 0,26 cm², Titanium, Opaque, 50 - 250 μl, PVDF, 1,0 µm, 96, -

Price
1 850,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particular laden samples. They are also widely used for clarification of aqueous solutions.

  • Multiple configurations and applications
  • Chemical compatibility
  • Automation-compatible
  • Superior vacuum filtration and filtrate collection
  • Hydrophilic lid
  • Colour code: Opaque

Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particulate-laden samples. They are also widely used for clarification of aqueous solutions.

Application:
Receptor/ligand binding assays, sample preparation, TCA precipitation of protein, DNA purification.
Research category and general filtration.
Nucleic acid and assays.

Binding type: High binding surface
Height: 14,4 mm
Length: 128 mm
Width: 85,5 mm

Specifications
  • Application:
    Receptor/ligand binding assays, sample preparation, enzyme assays with precipitation, DNA purification
  • Membrane:
    PVDF
  • No. of wells:
    96
  • Pore size:
    1,0 µm
  • Sterile:
    -
  • Colour:
    Opaque
  • Description:
    GFB plate, filtration area 0,26 cm²
  • Material:
    Titanium
  • Seq:
    01
  • Well volume:
    50 - 250 μl