Test kits, digital titration method
Sulphite reagent kit: iodate-iodide method, 4 - 400 mg/l, 100 Tests
HACH2272300
: Hach
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
- Pk:100 Tests
- Description:Sulphite reagent kit: iodate-iodide method
- Range:4 - 400 mg/l