Test kits, digital titration method
Hardness (Calcium) reagent set, high range, 10 - 4000 mg/l, 100 Tests
Catalog # HACH2447500
Supplier: Hach
Jump to:
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
- Pk:100 Tests
- Description:Hardness (Calcium) reagent set, high range
- Range:10 - 4000 mg/l