Test kits, digital titration method
Chloride reagent kit: Mercury nitrate method, 10 - 8000 mg/l, 100 Tests
Catalog # HACH2272600
Supplier: Hach
Jump to:
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
- Pk:100 Tests
- Description:Chloride reagent kit: Mercury nitrate method
- Range:10 - 8000 mg/l