Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
24/29 NS, 19/26 NS
Catalog # 201-8573
Supplier:   SciLabware
Multiple adapters, Quickfit®
Catalog # 201-8573
Supplier:  SciLabware
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
24/29 NS, 19/26 NS
Price
73,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, straight.
  • Both sockets are the same size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters with two parallel necks/sockets and a cone.
Spezifikationen
  • Cone size:
    24/29 NS
  • Socket size:
    19/26 NS
  • Seq:
    02