Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® I, M490

Tissue processing/embedding cassettes, Histosette® I, M490

Yellow
Catalog # 720-0890
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M490
Catalog # 720-0890
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M490
Tissue processing/embedding cassettes, Histosette® I, M490

Yellow

Price
163,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

Acetal polymer.

  • Resistant to histological solvents
  • Flow-through slots maximise fluid exchange and ensure proper drainage
  • Integral lid eliminates the need for separate steel lids
  • 30° writing angle

Documents

Specifications
  • Colour:
    Yellow
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.