Storage Boxes
Green, 595×395×320 mm
216-2879
PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.
- Very good chemical resistance
- Very good thermal resistance
- Very sturdy and economical
- Colour:Green
- Seq:15
- W×D×H:595×395×320 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



