Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Blue, 595×395×320 mm
Catalog # 216-2878
Supplier:   Gilac PROFESSIONNEL
Storage Boxes
Catalog # 216-2878
Storage Boxes
Storage Boxes

Blue, 595×395×320 mm

Price
148,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.

  • Very good chemical resistance
  • Very good thermal resistance
  • Very sturdy and economical
Specifications
  • Colour:
    Blue
  • Seq:
    16
  • W×D×H:
    595×395×320 mm