Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Green, 595×395×215 mm
Catalog # 216-2876
Supplier:   Gilac PROFESSIONNEL
Storage Boxes
Catalog # 216-2876
Storage Boxes
Storage Boxes

Green, 595×395×215 mm

Price
112,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.

  • Very good chemical resistance
  • Very good thermal resistance
  • Very sturdy and economical
Specifications
  • Colour:
    Green
  • Seq:
    13
  • W×D×H:
    595×395×215 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.