Storage Boxes
Green, 600×400×190 mm
Catalog # CORAG119711
Supplier: Gilac PROFESSIONNEL
Jump to:
Product Details & Documents
PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.
- Very good chemical resistance
- Very good thermal resistance
- Very sturdy and economical
Specifications
- Colour:Green
- Seq:10
- W×D×H:600×400×190 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



